Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dinitrobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. D269925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D269925G 25 g
D26995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D269925G Supplier TCI America Supplier No. D269925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 401-99-0
Molecular Formula C7H3F3N2O4
Molecular Weight (g/mol) 236.106
MDL Number MFCD00007233
InChI Key QZADIXWDDVQVKM-UHFFFAOYSA-N
PubChem CID 123060
IUPAC Name 1,3-dinitro-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point 50°C
Color Yellow
Boiling Point 129°C
Quantity 25 g
Synonym 3,5-dinitrobenzotrifluoride, 1,3-dinitro-5-trifluoromethyl benzene, benzene, 1,3-dinitro-5-trifluoromethyl, 3,5-dinitrotrifluorotoluene, alpha,alpha,alpha-trifluoro-3,5-dinitrotoluene, pubchem10396, 3, 5-dinitrotrifluorotoluene, 3,5-dinitro-benzotrifluoride, 3,5-?dinitrobenzotrifluoride
Formula Weight 236.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dinitrobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.