Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dinitrobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D414425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D414425G 25 g
D41445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D414425G Supplier TCI America Supplier No. D414425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71022-43-0
Molecular Formula C7H6N2O5
Molecular Weight (g/mol) 198.13
MDL Number MFCD00007235
InChI Key GPHYIQCSMDYRGJ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol #
PubChem CID 116978
IUPAC Name (3,5-dinitrophenyl)methanol
SMILES OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 91°C
Color Yellow
Quantity 25 g
Formula Weight 198.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dinitrobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.