Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dinitrosalicylic Acid, 98%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
609-99-4
C7H4N2O7
228.12
LWFUFLREGJMOIZ-UHFFFAOYSA-N
2-hydroxy-3,5-dinitrobenzoic acid
OC(=O)C1=CC(=CC(=C1O)[N+]([O-])=O)[N+]([O-])=O
609-99-4
C7H4N2O7
228.12
LWFUFLREGJMOIZ-UHFFFAOYSA-N
2-hydroxy-3,5-dinitrobenzoic acid
OC(=O)C1=CC(=CC(=C1O)[N+]([O-])=O)[N+]([O-])=O

D2288, 609-99-4, C7H4N2O7

Spectrum™ 3,5-Dinitrosalicylic Acid , also known as DNS or DNSA, is used for quantifying carbohydrates levels in blood. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Fiber Drum
Quantity 25 kg
CAS Max % 100%
Percent Purity 98%