Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyltrimethylammonium hydroxide, 0.1 M in methanol

Catalog No. AC422020025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1899-02-1 | C9H15NO | 153.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyltrimethylammonium hydroxide
Color Colorless
Name Note 0.1M in Methanol
CAS 1899-02-1, 67-56-1
Molecular Formula C9H15NO
Linear Formula C6H5N(CH3)3OH
MDL Number MFCD00041899
Concentration 0.09 to 0.11M
Density 0.8000g/mL
Synonym methelute, phenyltrimethylammonium hydroxide, trimethylanilinium hydroxide, phenyl trimethyl ammonium hydroxide, trimethylphenylammonium hydroxide, n,n,n-trimethylbenzenaminium hydroxide, n,n,n-trimethylanilinium hydroxide, benzenaminium, n,n,n-trimethyl-, hydroxide, ptah, ammonium, phenyltrimethyl-, hydroxide
InChI Key HADKRTWCOYPCPH-UHFFFAOYSA-M
SMILES [OH-].C[N+](C)(C)C1=CC=CC=C1
IUPAC Name N,N,N-trimethylanilinium hydroxide
Molecular Weight (g/mol) 153.23
PubChem CID 15913
ChEBI CHEBI:85062
Formula Weight 153.22
Packaging Plastic bottle
Specific Gravity 0.8
Quantity 2.5 kg
Flash Point 10°C
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.