Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Lutidine, 98+%

Catalog No. AC125215000
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle

CAS: 591-22-0 | C7H9N | 107.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-22-0
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.15
MDL Number MFCD00006404
InChI Key HWWYDZCSSYKIAD-UHFFFAOYSA-N
Synonym 3,5-lutidine, pyridine, 3,5-dimethyl, unii-6q4ypz045v, 3,5-dimethyl pyridine, 3,5 lutidine, 3,5-lutdcine, 3,5-dimethylpyridin, 3,5-dimethyl-pyridine, acmc-1axev, dsstox_cid_30825
PubChem CID 11565
IUPAC Name 3,5-dimethylpyridine
SMILES CC1=CC(=CN=C1)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -9.0°C
Color Yellow
Density 0.9300g/mL
Boiling Point 169.0°C to 170.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5040 to 1.5060
Quantity 500 mL
Beilstein 20, 246
Specific Gravity 0.93
Solubility Information Solubility in water: 33g/L (20°C). Other solubilities: soluble in ethanol and ether
Formula Weight 107.15
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3, 5-Lutidine, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Harmful in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 209-708-6
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.