Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(5-Methyl-1H-pyrazol-4-yl)propylamine, Technical Grade, Thermo Scientific™

Catalog No. SEW05905CB
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 28739-42-6
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.202
InChI Key BYGCQGLHPCERCH-UHFFFAOYSA-N
Synonym 3-5-methyl-1h-pyrazol-4-yl propan-1-amine, 3-5-methyl-1h-pyrazol-4-yl propylamine, 3-3-methyl-2h-pyrazol-4-yl propan-1-amine, 4-3-aminopropyl-5-methylpyrazole, 3-5-methylpyrazol-4-yl propylamine, 1h-pyrazole-4-propanamine,3-methyl, 4-3-aminopropyl-5-methyl-1h-pyrazole, tech
PubChem CID 2736943
IUPAC Name 3-(5-methyl-1H-pyrazol-4-yl)propan-1-amine
SMILES CC1=C(C=NN1)CCCN

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 139.2
Percent Purity 97%
Chemical Name or Material 3-(5-Methyl-1H-pyrazol-4-yl)propylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.