Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(5-Methyl-2-furyl)butyraldehyde, 99%

Catalog No. p-7026597
Click to view available options
Quantity:
5 g
25 g

CAS: 31704-80-0 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31704-80-0
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00209519
InChI Key PFLUFBMKOXFWSD-UHFFFAOYNA-N
Synonym 3-5-methyl-2-furyl butyraldehyde, 3-5-methylfuran-2-yl butanal, 3-5-methyl-2-furyl butanal, 3-5-methyl-2-furanyl butanal, fema no. 3307, beta5-dimethyl-2-furanpropanal, 2-furanpropanal, beta5-dimethyl, 2-furanpropanal, beta,5-dimethyl, beta5-dimethyl-2-furanpropionaldehyde, 2-furanpropanal, .beta.,5-dimethyl
PubChem CID 3578033
IUPAC Name 3-(5-methylfuran-2-yl)butanal
SMILES CCC(C=O)C1=CC=C(C)O1

Specifications

Density 1.006
Boiling Point 93°C to 94°C (14 mmHg)
Flash Point 58°C (136°F)
Quantity 5 g
UN Number UN1989
Sensitivity Air Sensitive
Formula Weight 152.19
Percent Purity 99%
Chemical Name or Material 3-(5-Methyl-2-furyl)butyraldehyde
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 250-771-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.