Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Pyrazoledicarboxylic acid monohydrate, 97%

Catalog No. p-188804
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 303180-11-2 | C5H4N2O4·H2O | 174.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303180-11-2
Molecular Formula C5H4N2O4·H2O
Molecular Weight (g/mol) 174.11
MDL Number MFCD00149323
InChI Key GLINCONFUZIMCN-UHFFFAOYSA-N
Synonym 3,5-pyrazoledicarboxylic acid, pyrazole-3,5-dicarboxylic acid, pyrazole-3,5-dicarboxylic acid monohydrate, 3,5-pyrazol dicarboxylic acid, 3,5-pyrazole dicarboxylic acid, zlchem 726, 3,5-dicarboxypyrazole, acmc-20a0hc, ksc223c6j, 3,5-pyrazole-dicarboxylic acid
PubChem CID 76559
SMILES O.OC(=O)C1=CC(=NN1)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 292°C to 295°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 25, 162
Solubility Information Solubility in water: insoluble
IUPAC Name 1H-pyrazole-3,5-dicarboxylic acid hydrate
Formula Weight 174.11
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Pyrazoledicarboxylic acid monohydrate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 221-474-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.