Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[3-(6-Aminohexylamino)propyl]trimethoxysilane 97.0+%, TCI America™
SDP

Catalog No. A312825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A312825ML 25 mL
A31285ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A312825ML Supplier TCI America Supplier No. A312825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51895-58-0
Molecular Formula C12H30N2O3Si
Molecular Weight (g/mol) 278.47
MDL Number MFCD00054943
InChI Key AMVXVPUHCLLJRE-UHFFFAOYSA-N
Synonym N-[3-(Trimethoxysilyl)propyl]hexamethylenediamine, N1-[3-(Trimethoxysilyl)propyl]hexane-1,6-diamine
PubChem CID 9925638
IUPAC Name (6-aminohexyl)[3-(trimethoxysilyl)propyl]amine
SMILES CO[Si](CCCNCCCCCCN)(OC)OC

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 mL
Formula Weight 278.47
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material [3-(6-Aminohexylamino)propyl]trimethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.