Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Di-tert-butylcarbazole 98.0+%, TCI America™
SDP

Catalog No. D39521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39521G 1 g
D39525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39521G Supplier TCI America Supplier No. D39521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37500-95-1
Molecular Formula C20H25N
Molecular Weight (g/mol) 279.43
MDL Number MFCD03425849
InChI Key OYFFSPILVQLRQA-UHFFFAOYSA-N
PubChem CID 15469209
IUPAC Name 3,6-di-tert-butyl-9H-carbazole
SMILES CC(C)(C)C1=CC2=C(NC3=C2C=C(C=C3)C(C)(C)C)C=C1

Specifications

Melting Point 232°C
Color White
Quantity 1 g
Formula Weight 279.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Di-tert-butylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.