Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dibromo-9-(2-ethylhexyl)carbazole 98.0+%, TCI America™
SDP

Catalog No. D48301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48301G 1 g
D48305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48301G Supplier TCI America Supplier No. D48301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 173063-52-0
Molecular Formula C20H23Br2N
Molecular Weight (g/mol) 437.22
MDL Number MFCD18252907
InChI Key ZDFZWZGIGKUBRA-UHFFFAOYNA-N
PubChem CID 16752909
IUPAC Name 3,6-dibromo-9-(2-ethylhexyl)-9H-carbazole
SMILES CCCCC(CC)CN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 437.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,6-Dibromo-9-(2-ethylhexyl)carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.