Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Diethynylcarbazole 99.0+%, TCI America™
SDP

Catalog No. D4275200MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg

Specifications

CAS 909342-65-0
Melting Point 199°C
Molecular Formula C16H9N
MDL Number MFCD24553219
Quantity 200 mg
InChI Key JNLNYORJQOYKHH-UHFFFAOYSA-N
SMILES C#CC1=CC2=C(NC3=C2C=C(C=C3)C#C)C=C1
IUPAC Name 3,6-diethynyl-9H-carbazole
Molecular Weight (g/mol) 215.26
PubChem CID 91659013
Formula Weight 215.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Diethynylcarbazole
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.