Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Diphenyl-1,2,4,5-tetrazine 98.0+%, TCI America™
SDP

Catalog No. D31751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31751G 1 g
D31755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31751G Supplier TCI America Supplier No. D31751G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6830-78-0
Molecular Formula C14H10N4
Molecular Weight (g/mol) 234.26
MDL Number MFCD00023646
InChI Key XAUWSIIGUUMHQQ-UHFFFAOYSA-N
Synonym diphenyl-s-tetrazine, 3,6-diphenyl-s-tetrazine, 3,6-diphenyltetrazine, 1,2,4,5-tetrazine, 3,6-diphenyl, s-tetrazine, 2,6-diphenyl, s-tetrazine, 3,6-diphenyl, 3,6-diphenyl-sym-tetrazine, diphenyl-1,2,4,5-tetrazine, acmc-1baef, enamine_004961
PubChem CID 23263
IUPAC Name diphenyl-1,2,4,5-tetrazine
SMILES C1=CC=C(C=C1)C1=NN=C(N=N1)C1=CC=CC=C1

Specifications

Melting Point 196°C
Quantity 1 g
Formula Weight 234.26
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Diphenyl-1,2,4,5-tetrazine
RTECSNumber XF6863300
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.