Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,7-Dithia-1,9-nonanediol 92.0+%, TCI America™
SDP

Catalog No. D284525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D284525G 25 g
D28455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D284525G Supplier TCI America Supplier No. D284525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16260-48-3
Molecular Formula C7H16O2S2
Molecular Weight (g/mol) 196.32
MDL Number MFCD01098524
InChI Key JIJWGPUMOGBVMQ-UHFFFAOYSA-N
PubChem CID 373735
IUPAC Name 2-({3-[(2-hydroxyethyl)sulfanyl]propyl}sulfanyl)ethan-1-ol
SMILES OCCSCCCSCCO

Specifications

Color Yellow
Quantity 25 g
Synonym 1,3-Bis(2-hydroxyethylthio)propane, 2,2′-(Trimethylenedithio)diethanol
Formula Weight 196.32
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,7-Dithia-1,9-nonanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.