Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,9'-Bicarbazole 98.0+%, TCI America™
SDP

Catalog No. B48111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48111G 1 g
B48115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48111G Supplier TCI America Supplier No. B48111G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18628-07-4
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.406
MDL Number MFCD27644690
InChI Key FHJJVSJWFYYPAC-UHFFFAOYSA-N
PubChem CID 13236022
IUPAC Name 3-carbazol-9-yl-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)N4C5=CC=CC=C5C6=CC=CC=C64

Specifications

Melting Point 216°C
Quantity 1 g
Formula Weight 332.41
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,9′-Bicarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.