Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane 98.0+%, TCI America™
SDP

Catalog No. D1027100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1027100G 100 g
D102725G 25 g
D1027500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1027100G Supplier TCI America Supplier No. D1027100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78-19-3
Molecular Formula C11H16O4
Molecular Weight (g/mol) 212.25
MDL Number MFCD00006564
InChI Key OOXMQACSWCZQLX-UHFFFAOYSA-N
Synonym 3,9-divinyl-2,4,8,10-tetraoxaspiro 5.5 undecane, 3,9-divinylspirobi m-dioxane, diallylidene pentaerythritol, diallylidenepentaerythritol, pentaerythritol diacrolein acetal, acrolein pentaerythritol bisacetal, acrolein, cyclic diacetal with pentaerythritol, acrolein-pentaerythritol dicyclic acetal, acrolein, cyclic neopentanetetrayl acetal, 2,4,8,10-tetraoxaspiro 5.5 undecane, 3,9-diethenyl
PubChem CID 66218
IUPAC Name 3,9-diethenyl-2,4,8,10-tetraoxaspiro[5.5]undecane
SMILES C=CC1OCC2(CO1)COC(OC2)C=C

Specifications

Melting Point 44°C
Color White
Boiling Point 94°C
Quantity 100 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane
EINECSNumber (5)-3770
RTECSNumber XF0875000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.