Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,9-Dodecadiyne 97.0+%, TCI America™
SDP

Catalog No. D19855ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL

Chemical Identifiers

CAS 61827-89-2
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00041637
InChI Key SLQXYOMUNKJHRV-UHFFFAOYSA-N
Synonym 3,9-dodecadiyne, acmc-209mwh, 3,9-dodecadiyne,
PubChem CID 143841
IUPAC Name dodeca-3,9-diyne
SMILES CCC#CCCCCC#CCC

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 162.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,9-Dodecadiyne
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.