Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetoxy-2-methylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A155825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A155825G 25 g
A15585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A155825G Supplier TCI America Supplier No. A155825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 168899-58-9
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00957176
InChI Key LHVJUPHCLWIPLZ-UHFFFAOYSA-N
Synonym 3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid
PubChem CID 3798557
IUPAC Name 3-acetyloxy-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1OC(=O)C)C(=O)O

Specifications

Melting Point 148°C
Color White
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetoxy-2-methylbenzoic Acid
EINECSNumber (3)-4420&(3)-4455
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.