Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetyl-2,4-dimethylpyrrole, 98%, Thermo Scientific Chemicals

Catalog No. p-4509359
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-25-6
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
InChI Key VGZCKCJMYREIKA-UHFFFAOYSA-N
Synonym 3-acetyl-2,4-dimethylpyrrole, 2,4-dimethyl-3-acetylpyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl ethanone, 2,4-dimethylpyrrol-3-yl methyl ketone, ethanone, 1-2,4-dimethyl-1h-pyrrol-3-yl, ketone, 2,4-dimethylpyrrol-3-yl methyl, 1-2,4-dimethyl-1h-pyrrol-3-yl ethan-1-one, 2,4-dimethyl-3-acetyl-pyrrole, 3-acetyl-2,5-dimethyl-pyrrole, 1-2,4-dimethyl-1h-pyrrol-3-yl-ethanone
PubChem CID 15163
IUPAC Name 1-(2,4-dimethyl-1H-pyrrol-3-yl)ethanone
SMILES CC1=CNC(=C1C(=O)C)C

Specifications

Melting Point 136.0°C to 139.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 137.18
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 3-Acetyl-2, 4-dimethylpyrrole
Hazard Category GHS Signal Word: Warning
EINECSNumber 219-197-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.