Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene 98.0+%, TCI America™
SDP

Catalog No. A26671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26671G 1 g
A26675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26671G Supplier TCI America Supplier No. A26671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 160982-10-5
Molecular Formula C6H6ClNO3S2
Molecular Weight (g/mol) 239.69
MDL Number MFCD09033309
InChI Key ODLFFSHLXVZFPY-UHFFFAOYSA-N
Synonym 3-Acetyl-5-chloro-2-thiophenesulfonamide
PubChem CID 15294528
IUPAC Name 3-acetyl-5-chlorothiophene-2-sulfonamide
SMILES CC(=O)C1=C(SC(Cl)=C1)S(N)(=O)=O

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 1 g
Formula Weight 239.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.