Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylbenzonitrile, 97%, Thermo Scientific™

Catalog No. AC307840050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6136-68-1
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001806
InChI Key SBCFGFDAZCTSRH-UHFFFAOYSA-N
Synonym m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649
PubChem CID 80222
IUPAC Name 3-acetylbenzonitrile
SMILES CC(=O)C1=CC=CC(=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 96.0°C to 100.0°C
Color Brown to Orange-Yellow
Boiling Point 120.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3COC6H4CN
Quantity 5 g
Formula Weight 145.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Acetylbenzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.