Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(alpha-Cyanobenzyl)-7-methyl-2(1H)-quinoxalinone, 97%, Thermo Scientific™

Catalog No. AAH5849503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5849503 1 g
AAH5849506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5849503 Supplier Thermo Scientific Chemicals Supplier No. H5849503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858240-50-2
Molecular Formula C17H13N3O
Molecular Weight (g/mol) 275.311
MDL Number MFCD22988956
InChI Key VBHNCMAFPOPJJC-UHFFFAOYSA-N
Synonym 2-6-methyl-3-oxo-4h-quinoxalin-2-yl-2-phenylacetonitrile, 3-alpha-cyanobenzyl-7-methyl-2 1h-quinoxalinone
PubChem CID 121235641
IUPAC Name 2-(6-methyl-3-oxo-4H-quinoxalin-2-yl)-2-phenylacetonitrile
SMILES CC1=CC2=C(C=C1)N=C(C(=O)N2)C(C#N)C3=CC=CC=C3

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 275.31
Percent Purity 97%
Chemical Name or Material 3-(alpha-Cyanobenzyl)-7-methyl-2(1H)-quinoxalinone
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.