Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-(2,4,6-trichlorophenyl)-5-pyrazolone , 97%, Thermo Scientific™

Catalog No. AAH6602203
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27241-31-2
Molecular Formula C9H6Cl3N3O
Molecular Weight (g/mol) 278.52
MDL Number MFCD00020743,MFCD11053342
InChI Key FOGGKKWSFIESMZ-UHFFFAOYSA-N
Synonym 3-amino-1-2,4,6-trichlorophenyl-1h-pyrazol-5 4h-one, 3-amino-1-2,4,6-trichlorophenyl-2-pyrazolin-5-one, 3-amino-1-2,4,6-trichlorophenyl-5-pyrazolone, 5-amino-2-2,4,6-trichlorophenyl-4h-pyrazol-3-one, 1-2,4,6-trichlorophenyl-3-anilinopyrazolone, 3h-pyrazol-3-one, 5-amino-2,4-dihydro-2-2,4,6-trichlorophenyl, 5-amino-2-2,4,6-trichlorophenyl-2,4-dihydro-3h-pyrazol-3-one, 1-2,4,6-trichlorophenyl 3-amino-5-pyrazolone, 3-amino-1-2,4,6-trichlorophenyl-4,5-dihydro-1h-pyrazol-5-one, 1-2,4,6-trichlorophenyl-3-amino-5-pyrazolone
PubChem CID 270746
IUPAC Name 3-amino-1-(2,4,6-trichlorophenyl)-4,5-dihydro-1H-pyrazol-5-one
SMILES NC1=NN(C(=O)C1)C1=C(Cl)C=C(Cl)C=C1Cl

Specifications

Quantity 1 g
Physical Form Solid
Beilstein 535761
Formula Weight 278.52
Percent Purity 97%
Chemical Name or Material 3-Amino-1-(2,4,6-trichlorophenyl)-5-pyrazolone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 248-358-9
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.