Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-propanesulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. A260225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A260225G 25 g
A26025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A260225G Supplier TCI America Supplier No. A260225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3687-18-1
Molecular Formula C3H9NO3S
Molecular Weight (g/mol) 139.17
MDL Number MFCD00008225
InChI Key SNKZJIOFVMKAOJ-UHFFFAOYSA-N
Synonym homotaurine, 3-amino-1-propanesulfonic acid, tramiprosate, alzhemed, 3-aminopropanesulphonic acid, 3aps, 1-propanesulfonic acid, 3-amino, cerebril, 3-aminopropanesulfonic acid, 3-aminopropane sulfonic acid
PubChem CID 1646
ChEBI CHEBI:1457
IUPAC Name 3-aminopropane-1-sulfonic acid
SMILES NCCCS(O)(=O)=O

Specifications

Melting Point 292°C
Color White
Quantity 25 g
Formula Weight 139.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-propanesulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.