Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1H-1,2,4-triazole, 96%

Catalog No. p-3344449
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 61-82-5 | C2H4N4 | 84.08 g/mol

3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.

Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-82-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00005230,MFCD00053362
InChI Key KLSJWNVTNUYHDU-UHFFFAOYSA-N
Synonym 3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
PubChem CID 1639
ChEBI CHEBI:40036
IUPAC Name 1H-1,2,4-triazol-5-amine
SMILES NC1=NC=NN1

Specifications

Melting Point 147°C to 153°C
Density 1.138
Assay Percent Range 96%
Quantity 50 g
UN Number UN3077
Beilstein 107687
Merck Index 14,489
Solubility Information It is soluble in water,methanol,ethanol and chloroform,and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.
Formula Weight 84.08
Percent Purity 96%
Chemical Name or Material 3-Amino-1, 2, 4-triazole
Show More Show Less
Hazard Category H302-H319-H350-H360-H373
Hazard Statement GHS H Statement
H361-H373
Precautionary Statement P201-P202-P260-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 200-521-5
RTECSNumber XZ3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.