Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-methylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. p-4380042
Click to view available options
Quantity:
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882678-96-8
Molecular Formula C13H20BNO2
Molecular Weight (g/mol) 233.12
MDL Number MFCD11054038
InChI Key JMKMGPGFYMANCA-UHFFFAOYSA-N
Synonym 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-2-methylphenylboronic acid, pinacol ester, 3-amino-2-methylphenylboronic acid pinacol ester, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine, 3-amino-2-methylphenyl boronic acid pinacol ester, 2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylamine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine
PubChem CID 46738005
IUPAC Name 2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1=C(C=CC=C1N)B1OC(C)(C)C(C)(C)O1

Specifications

Boiling Point 120°C to 121°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 233.12
Percent Purity 97%
Chemical Name or Material 3-Amino-2-methylphenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.