Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-(2-nitrophenyl)propionic acid, 98%

Catalog No. AAB2217606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2217606 5 g
AAB2217614 25 g
AAB2217622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2217606 Supplier Thermo Scientific Chemicals Supplier No. B2217606
Only null left
Add to Cart

CAS: 5678-48-8 | C9H10N2O4 | 210.189 g/mol

3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 0-5°C. Incompatible with strong oxidizing agents, acids, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5678-48-8
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.189
MDL Number MFCD00090356
InChI Key XXBOYULKNZTOMN-UHFFFAOYSA-N
Synonym 3-amino-3-2-nitrophenyl propanoic acid, 3-amino-3-2-nitrophenyl propionic acid, 3-amino-3-2-nitro-phenyl-propionic acid, 3-2-nitrophenyl-dl-beta-alanine, 3-2-nitrophenyl-beta-alanine, rs-3-amino-3-2-nitrophenyl-propionic acid, maybridge1_004875, dl-beta-2-nitrophenyl alanine, beta-amino-o-nitrohydrocinnamic acid, hydrocinnamic acid, beta-amino-o-nitro
PubChem CID 2779217
IUPAC Name 3-amino-3-(2-nitrophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)C(CC(=O)O)N)[N+](=O)[O-]

Specifications

Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material 3-Amino-3-(2-nitrophenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.