Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4,4,4-trifluorobutyric acid, 97%

Catalog No. AAL1223903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1223903 1 g
AAL12239MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1223903 Supplier Thermo Scientific Chemicals Supplier No. L1223903
Add to Cart

CAS: 584-20-3 | C4H6F3NO2 | 157.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-20-3
Molecular Formula C4H6F3NO2
Molecular Weight (g/mol) 157.09
MDL Number MFCD00041413
InChI Key XRXQOEWWPPJVII-REOHCLBHSA-N
PubChem CID 342970
IUPAC Name 3-amino-4,4,4-trifluorobutanoic acid
SMILES [NH3+][C@@H](CC([O-])=O)C(F)(F)F

Specifications

Melting Point ∽184°C (decomposition)
Beilstein 1764276
Synonym 3-amino-4,4,4-trifluorobutyric acid, butanoic acid, 3-amino-4,4,4-trifluoro, butanoic acid, 3-amino-4,4,4-trifluoro-, 3r, acmc-1ajx7, acmc-20n6c3, 3-trifluoromethyl-beta-alanine, butanoicacid,3-amino-4,4,4-trifluoro, butanoic acid,3-amino-4,4,4-trifluoro, r-3-amino-4,4,4-trifluorobutyric acid
Quantity 1 g
Formula Weight 157.09
Percent Purity 97%
Chemical Name or Material 3-Amino-4,4,4-trifluorobutyric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.