Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-bromo-1H-indazole, 95%, Thermo Scientific™

Catalog No. AC435790050
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 914311-50-5
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD09909580
InChI Key IXXXOIXGQFPLIH-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-1h-indazole, 4-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 4-bromo, 3-amino-4-bromoindazole, pubchem20681, acmc-209rbi, 4-bromo-1h-indazole-3-amine, 4-bromo-2h-indazol-3-ylamine, 4-bromo-1h-indazole-3-ylamine
PubChem CID 11975123
IUPAC Name 4-bromo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=CC(Br)=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 148.0°C to 150.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 212.05
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-bromo-1H-indazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.