Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-chlorobenzamide, 97%

Catalog No. aab2529406
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 19694-10-1 | C7H7ClN2O | 170.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19694-10-1
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD00035785
InChI Key QHMDKGRWJVOUFU-UHFFFAOYSA-N
PubChem CID 88200
IUPAC Name 3-amino-4-chlorobenzamide
SMILES NC(=O)C1=CC=C(Cl)C(N)=C1

Specifications

Melting Point 164°C to 166°C
Quantity 5 g
Synonym 3-amino-4-chloro-benzamide, benzamide, 3-amino-4-chloro, fast red base db-30, acmc-209f0q, 3-amino-4-chloro benzamide, benzamide,3-amino-4-chloro, 3-azanyl-4-chloranyl-benzamide, 3-amino-4-chlorobenzamide, 3-amino-4-chlorobenzoic acidamide
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 3-Amino-4-chlorobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-231-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.