Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-methoxybenzanilide 98.0+%, TCI America™
SDP

Catalog No. A1954250G
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 120-35-4
Molecular Formula C14H14N2O2
Molecular Weight (g/mol) 242.278
MDL Number MFCD00017166
InChI Key LHMQDVIHBXWNII-UHFFFAOYSA-N
Synonym 3-amino-4-methoxybenzanilide, 3-amino-p-anisanilide, benzamide, 3-amino-4-methoxy-n-phenyl, 3-amino-4-methoxy benzanilide, benzanilide, 3-amino-4-methoxy, p-anisanilide, 3-amino, unii-v5y646aw8s, amba, 3-amino-4-methoxy-n-phenyl-benzamide, 3-amino-4-methoxyphenyl-n-benzamide
PubChem CID 8426
IUPAC Name 3-amino-4-methoxy-N-phenylbenzamide
SMILES COC1=C(C=C(C=C1)C(=O)NC2=CC=CC=C2)N

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 250 g
Formula Weight 242.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methoxybenzanilide
EINECSNumber (3)-1554
RTECSNumber CV7400000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.