Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-methoxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. A095825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A095825G 25 g
A09585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A095825G Supplier TCI America Supplier No. A095825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39811-17-1
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD00007790
InChI Key DTYBRSLINXBXMP-UHFFFAOYSA-N
Synonym 5-phenyl-o-anisidine, 3-amino-4-methoxybiphenyl, 4-methoxy-1,1'-biphenyl-3-amine, 2-amino-4-phenylanisole, 4-methoxybiphenyl-3-amine, 1,1'-biphenyl-3-amine, 4-methoxy, 2-methoxy-5-phenylphenylamine, acmc-209j7y, 2-methoxy-5-phenyl-aniline, 3-amino-4-methoxy-biphenyl
PubChem CID 123489
IUPAC Name 2-methoxy-5-phenylaniline
SMILES COC1=C(C=C(C=C1)C2=CC=CC=C2)N

Specifications

Melting Point 84°C
Quantity 25 g
Formula Weight 199.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methoxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.