Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-methylbenzoic acid, 99%, Thermo Scientific™

Catalog No. AC103980050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 2458-12-0
MDL Number MFCD00007742
InChI Key XKFIFYROMAAUDL-UHFFFAOYSA-N
Synonym 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh
PubChem CID 75568
IUPAC Name 3-amino-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 164.0°C to 168.0°C
Color Gray to Pink
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H9NO2
Linear Formula H2NC6H3(CH3)CO2H
Quantity 5 g
Beilstein 14, 487
Molecular Weight (g/mol) 151.16
Formula Weight 151.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methylbenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-543-1
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.