Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Amino-4-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
| CAS | 22237-12-3 |
| Molecular Formula | C7H10BNO2 |
| Quantity | 5 g |
| MDL Number | MFCD01074640 |
| Synonym | 3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl |
| InChI Key | PLTGUDDQNWJILD-UHFFFAOYSA-N |
| SMILES | B(C1=CC(=C(C=C1)C)N)(O)O |
| IUPAC Name | (3-amino-4-methylphenyl)boronic acid |
| Molecular Weight (g/mol) | 150.972 |
| PubChem CID | 2737803 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction