Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-bromobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. A191425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A191425G 25 g
A19145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A191425G Supplier TCI America Supplier No. A191425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54962-75-3
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.023
InChI Key HJTLKVYOWNTDPF-UHFFFAOYSA-N
Synonym 3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine
PubChem CID 2735880
IUPAC Name 3-bromo-5-(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1N)Br)C(F)(F)F

Specifications

Boiling Point 223°C
Quantity 25 g
Formula Weight 240.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Amino-5-bromobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.