Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-cyclopropyl-1H-pyrazole, 96%, Thermo Scientific™

Catalog No. AAH5089803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5089803 1 g
AAH50898MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5089803 Supplier Thermo Scientific Chemicals Supplier No. H5089803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175137-46-9
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.159
MDL Number MFCD08461681
InChI Key MXVAGCQKBDMKPG-UHFFFAOYSA-N
Synonym 3-cyclopropyl-1h-pyrazol-5-amine, 3-amino-5-cyclopropyl-1h-pyrazole, 5-cyclopropyl-2h-pyrazol-3-ylamine, 3-amino-5-cyclopropylpyrazole, 1h-pyrazol-3-amine, 5-cyclopropyl, 3-cyclopropylpyrazol-5-amine, 5-cyclopropyl-2h-pyrazol-3-amine, 3-cyclopropyl-1h-pyrazole-5-amine, 5-cyclopropyl-1h-pyrazole-3-amine, 3-cyclopropylpyrazole-5-ylamine
PubChem CID 2758014
IUPAC Name 5-cyclopropyl-1H-pyrazol-3-amine
SMILES C1CC1C2=CC(=NN2)N

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 123.16
Percent Purity 96%
Chemical Name or Material 3-Amino-5-cyclopropyl-1H-pyrazole
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.