Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-phenylpyrazole, 98%, Thermo Scientific™

Catalog No. AC368020050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 1572-10-7
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00191749
InChI Key PWSZRRFDVPMZGM-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazol-5-amine, 3-amino-5-phenylpyrazole, 5-amino-3-phenylpyrazole, 1h-pyrazol-3-amine, 5-phenyl, 5-phenyl-2h-pyrazol-3-amine, pyrazole, 5-amino-3-phenyl, 3-amino-5-phenyl-1h-pyrazole, 5-phenyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-phenyl, 5-phenyl-1h-pyrazol-3-ylamine
PubChem CID 136655
IUPAC Name 5-phenyl-1H-pyrazol-3-amine
SMILES C1=CC=C(C=C1)C2=CC(=NN2)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 118°C to 121°C
Color Brown to Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 148
Formula Weight 159.19
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 3-Amino-5-phenylpyrazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.