Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Amino-6-chloropyridazine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 210.0°C to 211.0°C |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Formula Weight | 129.55 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Amino-6-chloropyridazine |
Safety and Handling
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
| EINECSNumber | 226-796-1 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction