Learn More
3-Amino-9-ethylcarbazole, 90%, tech.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 88.0 |
| CAS Max % | 100.0 |
| Melting Point | 98.0°C to 100.0°C |
| Assay Percent Range | 88% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Beilstein | 22, IV, 4936 |
| Solubility Information | Solubility in water: insoluble. |
| Formula Weight | 210.28 |
| Percent Purity | 90% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. May cause cancer. |
| Precautionary Statement | GHS P Statement Wash face,hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Obtain special instructions before use. Wear protective gloves/protective clothing/e |
| EINECSNumber | 205-057-7 |
| RTECSNumber | FE3590000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.