Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-9-ethylcarbazole, 90%, tech.

Catalog No. p-55483
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 132-32-1 | C14H14N2 | 210.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-32-1
Molecular Formula C14H14N2
Molecular Weight (g/mol) 210.28
MDL Number MFCD00004964
InChI Key OXEUETBFKVCRNP-UHFFFAOYSA-N
Synonym 3-amino-9-ethylcarbazole, 9-ethyl-9h-carbazol-3-amine, 3-amino-n-ethylcarbazole, 9h-carbazol-3-amine, 9-ethyl, carbazole, 3-amino-9-ethyl, 9-ethylcarbazol-3-ylamine, unii-8q2bg27jbu, 9-ethylcarbazole-3-ylamine, 8q2bg27jbu, 9-ethyl-9h-carbazol-3-ylamine
PubChem CID 8588
IUPAC Name 9-ethylcarbazol-3-amine
SMILES CCN1C2=C(C=C(C=C2)N)C3=CC=CC=C31

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 98.0°C to 100.0°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 22, IV, 4936
Solubility Information Solubility in water: insoluble.
Formula Weight 210.28
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Amino-9-ethylcarbazole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May cause cancer.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
Wear protective gloves/protective clothing/e
EINECSNumber 205-057-7
RTECSNumber FE3590000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.