Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzamide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3544-24-9
C7H8N2O
136.15
MFCD00007989
GSCPDZHWVNUUFI-UHFFFAOYSA-N
m-aminobenzamide,benzamide, 3-amino,3-amino-benzamide,benzamide, m-amino,meta-aminobenzamide,3-aba,3-aminobenzimide,3-amino benzamide,aniline-3-carboxamide,3-aminobenzoic acid amide
1645
CHEBI:64042
3-aminobenzamide
C1=CC(=CC(=C1)N)C(=O)N
3544-24-9
C7H8N2O
136.15
MFCD00007989
GSCPDZHWVNUUFI-UHFFFAOYSA-N
m-aminobenzamide,benzamide, 3-amino,3-amino-benzamide,benzamide, m-amino,meta-aminobenzamide,3-aba,3-aminobenzimide,3-amino benzamide,aniline-3-carboxamide,3-aminobenzoic acid amide
1645
CHEBI:64042
3-aminobenzamide
C1=CC(=CC(=C1)N)C(=O)N
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Beige
Melting Point 111.0°C to 115.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NC6H4CONH2
Solubility Information Solubility in water: soluble
Formula Weight 136.15
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3-Aminobenzamide
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 222-586-9