Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzamide, 98%

Catalog No. AC339080050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

Catalog No. AC339080050

Supplier: Thermo Scientific Chemicals 339080050

Only null left
Add to Cart

CAS: 3544-24-9 | C7H8N2O | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3544-24-9
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007989
InChI Key GSCPDZHWVNUUFI-UHFFFAOYSA-N
Synonym m-aminobenzamide, benzamide, 3-amino, 3-amino-benzamide, benzamide, m-amino, meta-aminobenzamide, 3-aba, 3-aminobenzimide, 3-amino benzamide, aniline-3-carboxamide, 3-aminobenzoic acid amide
PubChem CID 1645
ChEBI CHEBI:64042
IUPAC Name 3-aminobenzamide
SMILES NC(=O)C1=CC=CC(N)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 111.0°C to 115.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H4CONH2
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 136.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Aminobenzamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 222-586-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.