Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzamidine dihydrochloride hydrate, 98%, Thermo Scientific™

Catalog No. AC228075000
Click to view available options
Quantity:
500 mg
25 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37132-68-6
Molecular Formula C7H11Cl2N3
Molecular Weight (g/mol) 208.086
MDL Number MFCD03790003
InChI Key FKFMMMVFFYPIEE-UHFFFAOYSA-N
Synonym dihydrogen 3-aminobenzenecarboximidamide dichloride
PubChem CID 74764793
IUPAC Name 3-aminobenzenecarboximidamide;hydron;dichloride
SMILES [H+].[H+].C1=CC(=CC(=C1)N)C(=N)N.[Cl-].[Cl-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown or Pink-White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula 2HCl·xH2O
Quantity 500 mg
Formula Weight 208.09
Percent Purity ≥97.5% (on dry substance)
Physical Form Powder
Chemical Name or Material 3-Aminobenzamidine dihydrochloride hydrate, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-355-0
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.