Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzonitrile, 99%

Catalog No. p-55533
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 2237-30-1 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2237-30-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00007756
InChI Key NJXPYZHXZZCTNI-UHFFFAOYSA-N
Synonym 3-cyanoaniline, m-aminobenzonitrile, m-cyanoaniline, 3-amino-benzonitrile, 3-aminobenzonitril, benzonitrile, m-amino, m-anthranilonitrile, benzonitrile, 3-amino, unii-15v3cw939x, 3-amino benzonitrile
PubChem CID 16702
IUPAC Name 3-aminobenzonitrile
SMILES C1=CC(=CC(=C1)N)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 54.0°C
Color Brown
Boiling Point 288.0°C to 290.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H4CN
Quantity 5 g
Beilstein 14, 391
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ehtanol,ethyl ether,acetone
Formula Weight 118.14
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 3-Aminobenzonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POI
EINECSNumber 218-800-5
RTECSNumber DI2454000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.