Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzylamine 98.0+%, TCI America™
SDP

Catalog No. A143125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A143125G 25 g
A14315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A143125G Supplier TCI America Supplier No. A143125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4403-70-7
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00078355
InChI Key ZDBWYUOUYNQZBM-UHFFFAOYSA-N
Synonym 3-aminobenzylamine, 3-aminomethyl aniline, m-aminobenzylamine, benzenemethanamine, 3-amino, 3-aminomethyl-aniline, pubchem5802, alpha-amino-m-toluidine, 3-aminomethyl phenylamine, 3-aminophenyl methylamine
PubChem CID 4628831
IUPAC Name 3-(aminomethyl)aniline
SMILES NCC1=CC(N)=CC=C1

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 134°C
Quantity 25 g
UN Number 3259
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminobenzylamine
EINECSNumber (3)-3897
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.