Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminocrotononitrile 98.0+%, TCI America™
SDP

Catalog No. A0944100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0944100G 100 g
A094425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0944100G Supplier TCI America Supplier No. A0944100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1118-61-2
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00008071,MFCD00008071
InChI Key DELJOESCKJGFML-DUXPYHPUSA-N
Synonym 3-aminocrotononitrile, 2z-3-aminobut-2-enenitrile, 3-amino-2-butenenitrile, z-3-aminobut-2-enenitrile, beta-aminocrotononitrile, 3-amino-2-butenonitrile, 2-amino-1-propenecarbonitrile, 3-amino-crotononitrile, beta-amino-crotononitrile, 2z-3-amino-2-butenenitrile
PubChem CID 5325263
IUPAC Name (2E)-3-aminobut-2-enenitrile
SMILES C\C(N)=C/C#N

Specifications

Melting Point 75°C
Color Yellow
Boiling Point 120°C
Quantity 100 g
Formula Weight 82.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminocrotononitrile
EINECSNumber (2)-1526
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.