Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Aminomethyl)pyridine, 98+%

Catalog No. p-7027123
Click to view available options
Quantity:
25 g
100 g

CAS: 3731-52-0 | C6H8N2 | 108.144 g/mol

It finds its application as a rubefacient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a rubefacient.

Solubility
Fully miscible in water. Soluble in chloroform, ethyl acetate and methanol.

Notes
Air sensitive. Store away from air, carbon dioxide and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3731-52-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006412
InChI Key HDOUGSFASVGDCS-UHFFFAOYSA-N
Synonym 3-aminomethyl pyridine, 3-pyridinemethanamine, 3-picolylamine, picolamine, 3-pyridylmethylamine, 3-pyridinylmethanamine, 3-pyridinemethylamine, 3-aminomethylpyridine, pyridin-3-yl methanamine, 3-pyridylmethanamine
PubChem CID 31018
IUPAC Name pyridin-3-ylmethanamine
SMILES C1=CC(=CN=C1)CN

Specifications

Melting Point -21°C
Density 1.062
pH 11 to 12
Boiling Point 226°C
Flash Point 100°C (212°F)
Odor Amine-like
Refractive Index 1.551
Quantity 25 g
UN Number UN2735
Beilstein 108056
Sensitivity Air Sensitive
Solubility Information Fully miscible in water. Soluble in chloroform,ethyl acetate and methanol.
Formula Weight 108.14
Percent Purity ≥99%
Chemical Name or Material 3-(Aminomethyl)pyridine
Show More Show Less
Hazard Category H302+H312-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 223-091-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.