Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminooxetane, 95%

Catalog No. aah57248md
Click to view available options
Quantity:
250 mg
500 mg

CAS: 21635-88-1 | C3H7NO | 73.095 g/mol

It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine.

Solubility
Slightly soluble in water

Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21635-88-1
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.095
MDL Number MFCD08544363
InChI Key OJEOJUQOECNDND-UHFFFAOYSA-N
Synonym 3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam
PubChem CID 9833923
IUPAC Name oxetan-3-amine
SMILES C1C(CO1)N

Specifications

Color Colorless to Yellow
Density 1.05 g/mL
Boiling Point 99°C
Flash Point 24°C (75°F)
Quantity 250 mg
UN Number UN1993
Sensitivity Air sensitive
Solubility Information Slightly soluble in water
Formula Weight 73.09
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Aminooxetane
Show More Show Less
Hazard Category H226-H301-H318
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P310-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.