Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminooxetane, 95%

Catalog No. p-4812912
Click to view available options
Quantity:
250 mg
500 mg

CAS: 21635-88-1 | C3H7NO | 73.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21635-88-1
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.09
InChI Key OJEOJUQOECNDND-UHFFFAOYSA-N
Synonym 3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam
PubChem CID 9833923
IUPAC Name oxetan-3-amine
SMILES C1C(CO1)N

Specifications

Boiling Point 99°C
Flash Point 24°C
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 73.09
Percent Purity 95%
Chemical Name or Material 3-Aminooxetane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water f

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.