Learn More
3-Aminophenol, 98+%
Description
A chemical used in the preparation of inhibitors of mammalian carbonic anhydrase isoforms. It is also used in the preparation of quinoline tethered fluorescent carbon dots for regulated anticancer discovery. It is employed in the production of TB drugs, salicylic acid, amino stabilizer and developers. 3-Aminophenol has been used as a stabilizer of chlorine-containing thermoplastics, although its principal use is as an intermediate in the production of 4-amino-2-hydroxybenzoic acid this isomer is also employed as a hair colorant and as a coupler molecule in hair dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A chemical used in the preparation of inhibitors of mammalian carbonic anhydrase isoforms. It is also used in the preparation of quinoline tethered fluorescent carbon dots for regulated anticancer discovery. It is employed in the production of TB drugs, salicylic acid, amino stabilizer and developers. 3-Aminophenol has been used as a stabilizer of chlorine-containing thermoplastics, although its principal use is as an intermediate in the production of 4-amino-2-hydroxybenzoic acid this isomer is also employed as a hair colorant and as a coupler molecule in hair dyes.
Solubility
Soluble in water (35 mg/ml at 20°C), ether, alcohol, amyl alcohol, and benzene (slightly).
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications
Specifications
| Melting Point | 122°C to 126°C |
| Density | 0.99 |
| pH | 6.8 |
| Boiling Point | 164°C (11 mmHg) |
| Flash Point | 178°C (352°F) |
| Odor | Characteristic |
| Quantity | 5000 g |
| UN Number | UN2512 |
| Beilstein | 636059 |
| Merck Index | 14,460 |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H500 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMINOPHENOLS (O-;M-;P-) |
| EINECSNumber | 209-711-2 |
| RTECSNumber | SJ4900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.